N-Methyl-N-(2-piperidinylmethyl)-1-butanamine dihydrochloride

051671-2.5G
101896-578EA 838.46 USD
101896-578
N-Methyl-N-(2-piperidinylmethyl)-1-butanamine dihydrochloride
N-Methyl-N-(2-piperidinylmethyl)-1-butanamine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₆Cl₂N₂
MDL Number: MFCD13561723
CAS Number: 1219960-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR