1-(2-Piperidinylmethyl)-3-pyrrolidinol dihydrochloride

051670-2.5G
101896-574EA 341.21 USD
101896-574
1-(2-Piperidinylmethyl)-3-pyrrolidinol dihydrochloride
1-(2-Piperidinylmethyl)-3-pyrrolidinol dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₂Cl₂N₂O
MDL Number: MFCD13561722
CAS Number: 1220021-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR