4-(2-Piperidinylmethyl)morpholine dihydrochloride

051669-2.5G
101896-570EA 838.46 USD
101896-570
4-(2-Piperidinylmethyl)morpholine dihydrochloride
4-(2-Piperidinylmethyl)morpholine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₂Cl₂N₂O
MDL Number: MFCD13561721
CAS Number: 81310-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR