1-(2-Piperidinylmethyl)piperidine dihydrochloride

051668-2.5G
101896-566EA 838.46 USD
101896-566
1-(2-Piperidinylmethyl)piperidine dihydrochloride
1-(2-Piperidinylmethyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂
MDL Number: MFCD13561720
CAS Number: 81310-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR