2-[Methyl(2-piperidinylmethyl)amino]-1-ethanol dihydrochloride

051667-2.5G
101896-562EA 838.46 USD
101896-562
2-[Methyl(2-piperidinylmethyl)amino]-1-ethanol dihydrochloride
2-[Methyl(2-piperidinylmethyl)amino]-1-ethanol dihydrochloride

Hazard: Irritant

Formula: C₉H₂₂Cl₂N₂O
MDL Number: MFCD13561719
CAS Number: 1220035-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR