1-(3-Pyrrolidinylmethyl)indoline dihydrochloride

051661-2.5G
101896-538EA 838.46 USD
101896-538
1-(3-Pyrrolidinylmethyl)indoline dihydrochloride
1-(3-Pyrrolidinylmethyl)indoline dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₀Cl₂N₂
MDL Number: MFCD13561713
CAS Number: 1220018-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR