N-Ethyl-N-(3-pyrrolidinylmethyl)cyclohexanamine dihydrochloride

051658-2.5G
101896-526EA 838.46 USD
101896-526
N-Ethyl-N-(3-pyrrolidinylmethyl)cyclohexanamine dihydrochloride
N-Ethyl-N-(3-pyrrolidinylmethyl)cyclohexanamine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₈Cl₂N₂
MDL Number: MFCD13561710
CAS Number: 1219964-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR