1-(3-Pyrrolidinylmethyl)-4-piperidinol dihydrochloride

051653-2.5G
101896-506EA 838.46 USD
101896-506
1-(3-Pyrrolidinylmethyl)-4-piperidinol dihydrochloride
1-(3-Pyrrolidinylmethyl)-4-piperidinol dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₂Cl₂N₂O
MDL Number: MFCD13561705
CAS Number: 1220027-06-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR