4-(3-Pyrrolidinylmethyl)-2-piperazinone dihydrochloride

051652-2.5G
101896-502EA 341.21 USD
101896-502
4-(3-Pyrrolidinylmethyl)-2-piperazinone dihydrochloride
4-(3-Pyrrolidinylmethyl)-2-piperazinone dihydrochloride

Hazard: Irritant

Formula: C₉H₁₉Cl₂N₃O
MDL Number: MFCD13561704
CAS Number: 1220021-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR