1-(3-Pyrrolidinylmethyl)azepane dihydrochloride

051651-2.5G
101896-498EA 838.46 USD
101896-498
1-(3-Pyrrolidinylmethyl)azepane dihydrochloride
1-(3-Pyrrolidinylmethyl)azepane dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂
MDL Number: MFCD13561703
CAS Number: 1219981-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR