N-Methyl-N-(3-pyrrolidinylmethyl)-1-butanamine dihydrochloride

051643-2.5G
101896-466EA 838.46 USD
101896-466
N-Methyl-N-(3-pyrrolidinylmethyl)-1-butanamine dihydrochloride
N-Methyl-N-(3-pyrrolidinylmethyl)-1-butanamine dihydrochloride

Hazard: Irritant

Formula: C₁₀H₂₄Cl₂N₂
MDL Number: MFCD13561695
CAS Number: 1219964-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR