N-Ethyl-N-(3-pyrrolidinylmethyl)-1-ethanamine dihydrochloride

051638-2.5G
101896-446EA 887.78 USD
101896-446
N-Ethyl-N-(3-pyrrolidinylmethyl)-1-ethanamine dihydrochloride
N-Ethyl-N-(3-pyrrolidinylmethyl)-1-ethanamine dihydrochloride

Hazard: Irritant

Formula: C₉H₂₂Cl₂N₂
MDL Number: MFCD13561690
CAS Number: 1219981-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR