N-Methyl-2-(4-piperidinyl)-N-(tetrahydro-2H-pyran-4-ylmethyl)-1-ethanamine dihydrochloride

051635-2.5G
101896-434EA 1061.78 USD
101896-434
N-Methyl-2-(4-piperidinyl)-N-(tetrahydro-2H-pyran-4-ylmethyl)-1-ethanamine dihydrochloride
N-Methyl-2-(4-piperidinyl)-N-(tetrahydro-2H-pyran-4-ylmethyl)-1-ethanamine dihydrochloride

Hazard: Irritant

Formula: C₁₄H₃₀Cl₂N₂O
MDL Number: MFCD13561687
CAS Number: 1219957-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR