N-Benzyl-N-methyl-2-(4-piperidinyl)-1-ethanamine dihydrochloride

Supplier: Matrix Scientific

051628-500MG
101896-404EA 364.62 USD
101896-404
N-Benzyl-N-methyl-2-(4-piperidinyl)-1-ethanamine dihydrochloride
N-Benzyl-N-methyl-2-(4-piperidinyl)-1-ethanamine dihydrochloride

Hazard: Irritant

Formula: C₁₅H₂₆Cl₂N₂
MDL Number: MFCD13561680
CAS Number: 1219961-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR