N-Methyl-N-[2-(4-piperidinyl)ethyl]-cyclohexanamine dihydrochloride

Supplier: Matrix Scientific

051626-500MG
101896-396EA 364.62 USD
101896-396
N-Methyl-N-[2-(4-piperidinyl)ethyl]-cyclohexanamine dihydrochloride
N-Methyl-N-[2-(4-piperidinyl)ethyl]-cyclohexanamine dihydrochloride

Hazard: Irritant

Formula: C₁₄H₃₀Cl₂N₂
MDL Number: MFCD13561678
CAS Number: 1219964-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR