1-[2-(4-Piperidinyl)ethyl]-4-piperidinol dihydrochloride

Supplier: Matrix Scientific

051624-500MG
101896-388EA 364.62 USD
101896-388
1-[2-(4-Piperidinyl)ethyl]-4-piperidinol dihydrochloride
1-[2-(4-Piperidinyl)ethyl]-4-piperidinol dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₆Cl₂N₂O
MDL Number: MFCD13561676
CAS Number: 1220038-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR