4-[2-(4-Piperidinyl)ethyl]-2-piperazinone dihydrochloride

Supplier: Matrix Scientific

051623-500MG
101896-384EA 364.62 USD
101896-384
4-[2-(4-Piperidinyl)ethyl]-2-piperazinone dihydrochloride
4-[2-(4-Piperidinyl)ethyl]-2-piperazinone dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₃Cl₂N₃O
MDL Number: MFCD13561675
CAS Number: 1219957-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR