1-[2-(4-Piperidinyl)ethyl]azepane dihydrochloride

Supplier: Matrix Scientific

051622-500MG
101896-380EA 272.55 USD
101896-380
1-[2-(4-Piperidinyl)ethyl]azepane dihydrochloride
1-[2-(4-Piperidinyl)ethyl]azepane dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₈Cl₂N₂
MDL Number: MFCD13561674
CAS Number: 1220035-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR