1-Benzyl-4-[2-(4-piperidinyl)ethyl]piperazine dihydrochloride

Supplier: Matrix Scientific

051621-500MG
101896-376EA 186.15 USD
101896-376
1-Benzyl-4-[2-(4-piperidinyl)ethyl]piperazine dihydrochloride
1-Benzyl-4-[2-(4-piperidinyl)ethyl]piperazine dihydrochloride

Hazard: Irritant

Formula: C₁₈H₃₁Cl₂N₃
MDL Number: MFCD13561673
CAS Number: 1220038-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR