1-Methyl-4-[2-(4-piperidinyl)ethyl]piperazine dihydrochloride

Supplier: Matrix Scientific

051620-500MG
101896-372EA 272.55 USD
101896-372
1-Methyl-4-[2-(4-piperidinyl)ethyl]piperazine dihydrochloride
1-Methyl-4-[2-(4-piperidinyl)ethyl]piperazine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₇Cl₂N₃
MDL Number: MFCD13561672
CAS Number: 1219957-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR