4-Methyl-1-[2-(4-piperidinyl)ethyl]piperidine dihydrochloride

Supplier: Matrix Scientific

051619-500MG
101896-368EA 364.62 USD
101896-368
4-Methyl-1-[2-(4-piperidinyl)ethyl]piperidine dihydrochloride
4-Methyl-1-[2-(4-piperidinyl)ethyl]piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₈Cl₂N₂
MDL Number: MFCD13561671
CAS Number: 30131-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR