2-Methyl-1-[2-(4-piperidinyl)ethyl]piperidine dihydrochloride

Supplier: Matrix Scientific

051617-500MG
101896-360EA 364.62 USD
101896-360
2-Methyl-1-[2-(4-piperidinyl)ethyl]piperidine dihydrochloride
2-Methyl-1-[2-(4-piperidinyl)ethyl]piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₈Cl₂N₂
MDL Number: MFCD13561669
CAS Number: 1219981-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR