1-[2-(4-Piperidinyl)ethyl]piperidine dihydrochloride

Supplier: Matrix Scientific

051612-500MG
101896-340EA 272.55 USD
101896-340
1-[2-(4-Piperidinyl)ethyl]piperidine dihydrochloride
1-[2-(4-Piperidinyl)ethyl]piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₆Cl₂N₂
MDL Number: MFCD13561664
CAS Number: 105903-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR