4-(2-(Pyrrolidin-1-yl)ethyl)piperidine dihydrochloride

Supplier: Matrix Scientific

Synonyms: 1-[2-(Piperidin-4-yl)ethyl]pyrrolidine dihydrochloride4-[2-(1-Pyrrolidinyl)ethyl]piperidine dihydrochloride

051609-500MG
101896-328EA 364.62 USD
101896-328
4-(2-(Pyrrolidin-1-yl)ethyl)piperidine dihydrochloride
4-(2-(Pyrrolidin-1-yl)ethyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂
MW: 255.23 g/mol
MDL Number: MFCD12974872
CAS Number: 14759-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR