1-[2-(3-Piperidinyl)ethyl]-1,2,3,4-tetrahydroquinoline dihydrochloride

Supplier: Matrix Scientific

051605-500MG
101896-312EA 139.15 USD
101896-312
1-[2-(3-Piperidinyl)ethyl]-1,2,3,4-tetrahydroquinoline dihydrochloride
1-[2-(3-Piperidinyl)ethyl]-1,2,3,4-tetrahydroquinoline dihydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆Cl₂N₂
MDL Number: MFCD13561659
CAS Number: 1219957-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR