2-{4-[2-(3-Piperidinyl)ethyl]-1-piperazinyl}-1-ethanol dihydrochloride

Supplier: Matrix Scientific

051602-500MG
101896-300EA 272.55 USD
101896-300
2-{4-[2-(3-Piperidinyl)ethyl]-1-piperazinyl}-1-ethanol dihydrochloride
2-{4-[2-(3-Piperidinyl)ethyl]-1-piperazinyl}-1-ethanol dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₉Cl₂N₃O
MDL Number: MFCD13561656
CAS Number: 1220038-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR