2-Ethyl-1-[2-(3-piperidinyl)ethyl]piperidine dihydrochloride

Supplier: Matrix Scientific

051599-500MG
101896-288EA 364.62 USD
101896-288
2-Ethyl-1-[2-(3-piperidinyl)ethyl]piperidine dihydrochloride
2-Ethyl-1-[2-(3-piperidinyl)ethyl]piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₄H₃₀Cl₂N₂
MDL Number: MFCD13561653
CAS Number: 1219981-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR