N-Methyl-N-[2-(3-piperidinyl)ethyl]-cyclohexanamine dihydrochloride

Supplier: Matrix Scientific

051598-500MG
101896-284EA 364.62 USD
101896-284
N-Methyl-N-[2-(3-piperidinyl)ethyl]-cyclohexanamine dihydrochloride
N-Methyl-N-[2-(3-piperidinyl)ethyl]-cyclohexanamine dihydrochloride

Hazard: Irritant

Formula: C₁₄H₃₀Cl₂N₂
MDL Number: MFCD13561652
CAS Number: 1219960-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR