N-[2-(3-Piperidinyl)ethyl]-N-propyl-1-propanamine dihydrochloride

Supplier: Matrix Scientific

051597-500MG
101896-280EA 364.62 USD
101896-280
N-[2-(3-Piperidinyl)ethyl]-N-propyl-1-propanamine dihydrochloride
N-[2-(3-Piperidinyl)ethyl]-N-propyl-1-propanamine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₃₀Cl₂N₂
MDL Number: MFCD13561651
CAS Number: 1220035-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR