1-[2-(3-Piperidinyl)ethyl]azepane dihydrochloride

Supplier: Matrix Scientific

051594-500MG
101896-268EA 364.62 USD
101896-268
1-[2-(3-Piperidinyl)ethyl]azepane dihydrochloride
1-[2-(3-Piperidinyl)ethyl]azepane dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₈Cl₂N₂
MDL Number: MFCD13561648
CAS Number: 1220018-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR