3-Methyl-1-[2-(3-piperidinyl)ethyl]piperidine dihydrochloride
Supplier: Matrix Scientific
051590-500MG
101896-252EA
364.62
USD
101896-252
3-Methyl-1-[2-(3-piperidinyl)ethyl]piperidine dihydrochloride
3-Methyl-1-[2-(3-piperidinyl)ethyl]piperidine dihydrochloride
Hazard: Irritant
Formula:
C₁₃H₂₈Cl₂N₂ |
MDL Number:
MFCD13561644 CAS Number: 1220030-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...