N-Allyl-N-[2-(3-piperidinyl)ethyl]-2-propen-1-amine dihydrochloride

Supplier: Matrix Scientific

051588-500MG
101896-244EA 364.62 USD
101896-244
N-Allyl-N-[2-(3-piperidinyl)ethyl]-2-propen-1-amine dihydrochloride
N-Allyl-N-[2-(3-piperidinyl)ethyl]-2-propen-1-amine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₆Cl₂N₂
MDL Number: MFCD13561642
CAS Number: 1219960-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR