1-[2-(3-Piperidinyl)ethyl]-3-pyrrolidinol dihydrochloride

Supplier: Matrix Scientific

051585-500MG
101896-232EA 364.62 USD
101896-232
1-[2-(3-Piperidinyl)ethyl]-3-pyrrolidinol dihydrochloride
1-[2-(3-Piperidinyl)ethyl]-3-pyrrolidinol dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂O
MDL Number: MFCD13561639
CAS Number: 1220034-53-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR