4-[2-(3-Piperidinyl)ethyl]morpholine dihydrochloride

Supplier: Matrix Scientific

051584-500MG
101896-228EA 364.62 USD
101896-228
4-[2-(3-Piperidinyl)ethyl]morpholine dihydrochloride
4-[2-(3-Piperidinyl)ethyl]morpholine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂O
MDL Number: MFCD13561638
CAS Number: 1219960-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR