1-[2-(3-Piperidinyl)ethyl]piperidine dihydrochloride

Supplier: Matrix Scientific

051583-500MG
101896-224EA 272.55 USD
101896-224
1-[2-(3-Piperidinyl)ethyl]piperidine dihydrochloride
1-[2-(3-Piperidinyl)ethyl]piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₆Cl₂N₂
MDL Number: MFCD13561637
CAS Number: 1220017-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR