N,N-Diethyl-2-(3-piperidinyl)-1-ethanamine dihydrochloride

Supplier: Matrix Scientific

051581-500MG
101896-216EA 272.55 USD
101896-216
N,N-Diethyl-2-(3-piperidinyl)-1-ethanamine dihydrochloride
N,N-Diethyl-2-(3-piperidinyl)-1-ethanamine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₆Cl₂N₂
MDL Number: MFCD13561635
CAS Number: 1220030-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR