1-[2-(2-Piperidinyl)ethyl]indoline dihydrochloride

051575-2.5G
101896-194EA 1023.41 USD
101896-194
1-[2-(2-Piperidinyl)ethyl]indoline dihydrochloride
1-[2-(2-Piperidinyl)ethyl]indoline dihydrochloride

Hazard: Irritant

Formula: C₁₅H₂₄Cl₂N₂
MDL Number: MFCD13561629
CAS Number: 1220034-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR