2-[2-(2-Piperidinyl)ethyl]-1,2,3,4-tetrahydroisoquinoline dihydrochloride

051574-2.5G
101896-190EA 838.46 USD
101896-190
2-[2-(2-Piperidinyl)ethyl]-1,2,3,4-tetrahydroisoquinoline dihydrochloride
2-[2-(2-Piperidinyl)ethyl]-1,2,3,4-tetrahydroisoquinoline dihydrochloride

Hazard: Irritant

Formula: C₁₆H₂₆Cl₂N₂
MDL Number: MFCD13561628
CAS Number: 1219981-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR