2-Ethyl-1-[2-(2-piperidinyl)ethyl]piperidine dihydrochloride

051570-2.5G
101896-174EA 838.46 USD
101896-174
2-Ethyl-1-[2-(2-piperidinyl)ethyl]piperidine dihydrochloride
2-Ethyl-1-[2-(2-piperidinyl)ethyl]piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₄H₃₀Cl₂N₂
MDL Number: MFCD13561624
CAS Number: 1219964-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR