N-[2-(2-Piperidinyl)ethyl]-N-propyl-1-propanamine dihydrochloride

051568-2.5G
101896-166EA 838.46 USD
101896-166
N-[2-(2-Piperidinyl)ethyl]-N-propyl-1-propanamine dihydrochloride
N-[2-(2-Piperidinyl)ethyl]-N-propyl-1-propanamine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₃₀Cl₂N₂
MDL Number: MFCD13561622
CAS Number: 1219963-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR