4-[2-(2-Piperidinyl)ethyl]-2-piperazinone dihydrochloride

051566-2.5G
101896-158EA 838.46 USD
101896-158
4-[2-(2-Piperidinyl)ethyl]-2-piperazinone dihydrochloride
4-[2-(2-Piperidinyl)ethyl]-2-piperazinone dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₃Cl₂N₃O
MDL Number: MFCD13561620
CAS Number: 1219960-59-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR