1-Methyl-4-[2-(2-piperidinyl)ethyl]piperazine dihydrochloride

051563-2.5G
101896-146EA 838.46 USD
101896-146
1-Methyl-4-[2-(2-piperidinyl)ethyl]piperazine dihydrochloride
1-Methyl-4-[2-(2-piperidinyl)ethyl]piperazine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₇Cl₂N₃
MDL Number: MFCD13561617
CAS Number: 1220021-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR