4-Methyl-1-[2-(2-piperidinyl)ethyl]piperidine dihydrochloride

051562-2.5G
101896-142EA 838.46 USD
101896-142
4-Methyl-1-[2-(2-piperidinyl)ethyl]piperidine dihydrochloride
4-Methyl-1-[2-(2-piperidinyl)ethyl]piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₈Cl₂N₂
MDL Number: MFCD13561616
CAS Number: 1219980-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR