3-Methyl-1-[2-(2-piperidinyl)ethyl]piperidine dihydrochloride

051561-2.5G
101896-138EA 838.46 USD
101896-138
3-Methyl-1-[2-(2-piperidinyl)ethyl]piperidine dihydrochloride
3-Methyl-1-[2-(2-piperidinyl)ethyl]piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₃H₂₈Cl₂N₂
MDL Number: MFCD13561615
CAS Number: 1219957-51-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR