N-Allyl-N-[2-(2-piperidinyl)ethyl]-2-propen-1-amine dihydrochloride
051559-2.5G
101896-130EA
838.46
USD
101896-130
N-Allyl-N-[2-(2-piperidinyl)ethyl]-2-propen-1-amine dihydrochloride
N-Allyl-N-[2-(2-piperidinyl)ethyl]-2-propen-1-amine dihydrochloride
Hazard: Irritant
Formula:
C₁₃H₂₆Cl₂N₂ |
MDL Number:
MFCD13561613 CAS Number: 1220021-44-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...