N-Cyclohexyl-N-ethyl-3-pyrrolidinamine dihydrochloride

051543-2.5G
101896-066EA 838.46 USD
101896-066
N-Cyclohexyl-N-ethyl-3-pyrrolidinamine dihydrochloride
N-Cyclohexyl-N-ethyl-3-pyrrolidinamine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₆Cl₂N₂
MDL Number: MFCD13561600
CAS Number: 1219980-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR