2-Ethyl-1-(3-pyrrolidinyl)piperidine dihydrochloride

051541-2.5G
101896-058EA 838.46 USD
101896-058
2-Ethyl-1-(3-pyrrolidinyl)piperidine dihydrochloride
2-Ethyl-1-(3-pyrrolidinyl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂
MDL Number: MFCD13561598
CAS Number: 1219957-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR