N-Cyclohexyl-N-methyl-3-pyrrolidinamine dihydrochloride

051540-2.5G
101896-054EA 838.46 USD
101896-054
N-Cyclohexyl-N-methyl-3-pyrrolidinamine dihydrochloride
N-Cyclohexyl-N-methyl-3-pyrrolidinamine dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₄Cl₂N₂
MDL Number: MFCD13561597
CAS Number: 1219957-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR