4-(3-Pyrrolidinyl)-2-piperazinone dihydrochloride

051537-2.5G
101896-042EA 838.46 USD
101896-042
4-(3-Pyrrolidinyl)-2-piperazinone dihydrochloride
4-(3-Pyrrolidinyl)-2-piperazinone dihydrochloride

Hazard: Irritant

Formula: C₈H₁₇Cl₂N₃O
MDL Number: MFCD13561594
CAS Number: 1219957-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR