N,N-Diallyl-3-pyrrolidinamine dihydrochloride
Supplier: Matrix Scientific
051530-2.5G
051530-500MG
101896-014EA
838.46
USD
101896-014
101896-012
N,N-Diallyl-3-pyrrolidinamine dihydrochloride
N,N-Diallyl-3-pyrrolidinamine dihydrochloride
Hazard: Irritant
Formula:
C₁₀H₂₀Cl₂N₂ |
MDL Number:
MFCD13561587 CAS Number: 1220020-24-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...